2-Bromo-3'-(trifluoromethyl)acetophenone 98%
|
|
BLDPBD81070-10GEA
137
EUR
BLDPBD81070-10G
BLDPBD81070-5G
BLDPBD81070-1G
BLDPBD81070-25G
2-Bromo-3'-(trifluoromethyl)acetophenone 98%
2-Bromo-3'-(trifluoromethyl)acetophenone
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₉H₆BrF₃O MW: 267,04 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD03094283 Núm. CAS: 2003-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...