3-(Methylthio)-1-hexanol 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD80757-5GEA 15 EUR
BLDPBD80757-5G BLDPBD80757-25G BLDPBD80757-100G
3-(Methylthio)-1-hexanol 98%
3-(Methylthio)-1-hexanol

Storage condition: Sealed in dry, Room Temperature

Formula: C₄H₁₀OS
MW: 148,26 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00010674
Núm. CAS: 51755-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR