3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate 95% estabilizado
|
|
Estabilización:
Stabilised with 4-methoxyphenol (MEHQ)
BLDPBD80720-100GEA
94
EUR
BLDPBD80720-100G
BLDPBD80720-500G
BLDPBD80720-25G
BLDPBD80720-5G
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate 95% estabilizado
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₆H₃₈O₅Si₄ MW: 422,82 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00053871 Núm. CAS: 17096-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...