3-(Butylcarbamoyl)phenylboronic acid 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD80516-5GEA 146 EUR
BLDPBD80516-5G BLDPBD80516-0.025G BLDPBD80516-1G BLDPBD80516-25G BLDPBD80516-250MG
3-(Butylcarbamoyl)phenylboronic acid 98%
3-(Butylcarbamoyl)phenylboronic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₆BNO₃
MW: 221,06 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD04115691
Núm. CAS: 397843-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR