3-(4-Methoxyphenyl)-5-isoxazolecarboxylic acid 95%

Proveedor: BLD PHARMATECH GMBH

BLDPBD80189-10GEA 306 EUR
BLDPBD80189-10G BLDPBD80189-1G BLDPBD80189-250MG BLDPBD80189-5G
3-(4-Methoxyphenyl)-5-isoxazolecarboxylic acid 95%
3-(4-Methoxyphenyl)-5-isoxazolecarboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₁H₉NO₄
MW: 219,1900 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD04124288
Núm. CAS: 618383-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR