Ethyl-3-(3-boronobenzamido)propionate 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD79710-250MGEA 66 EUR
BLDPBD79710-250MG BLDPBD79710-1G
Ethyl-3-(3-boronobenzamido)propionate 98%
Ethyl-3-(3-boronobenzamido)propionate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₂H₁₆BNO₅
MW: 265,07 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD04115701
Núm. CAS: 850567-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR