5-Amino-3-(3-bromophenyl)isoxazole 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD79667-100MGEA 39 EUR
BLDPBD79667-100MG
5-Amino-3-(3-bromophenyl)isoxazole 98%
5-Amino-3-(3-bromophenyl)isoxazole

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₉H₇BrN₂O
MW: 239,0687 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD07787076
Núm. CAS: 119162-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR