p-Toluenesulphonylacetonitrile 97%
|
|
BLDPBD7792-25GEA
58
EUR
BLDPBD7792-25G
BLDPBD7792-10G
BLDPBD7792-5G
BLDPBD7792-1G
p-Toluenesulphonylacetonitrile 97%
p-Toluenesulphonylacetonitrile
Storage condition: Sealed in dry, Room Temperature
Formula:
C₉H₉NO₂S MW: 195,24 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00039487 Núm. CAS: 5697-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...