2-Nitro-5-picoline 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD77657-250MGEA 7 EUR
BLDPBD77657-250MG BLDPBD77657-25G BLDPBD77657-100G BLDPBD77657-5G BLDPBD77657-1G BLDPBD77657-10G
2-Nitro-5-picoline 98%
2-Nitro-5-picoline

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₆N₂O₂
MW: 138,1200 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00234182
Núm. CAS: 1074-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR