3,5-Difluoro-DL-phenylalanine 97%

BLDPBD7713-5GEA 101 EUR
BLDPBD7713-5G BLDPBD7713-100MG BLDPBD7713-250MG BLDPBD7713-1G
3,5-Difluoro-DL-phenylalanine 97%
3,5-Difluoro-DL-phenylalanine

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₉F₂NO₂
MW: 201,1701 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00061308
Núm. CAS: 32133-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR