5'-Chloro-2'-methoxyacetophenone 95%

Proveedor: BLD PHARMATECH GMBH

BLDPBD7701-25GEA 708 EUR
BLDPBD7701-25G BLDPBD7701-1G BLDPBD7701-100G BLDPBD7701-250MG BLDPBD7701-5G
5'-Chloro-2'-methoxyacetophenone 95%
5'-Chloro-2'-methoxyacetophenone

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₉ClO₂
MW: 184,6196 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00174339
Núm. CAS: 6342-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR