2-Bromo-4'-(methylsulphonyl)acetophenone 98%
|
|
BLDPBD7679-1GEA
15
EUR
BLDPBD7679-1G
BLDPBD7679-0.025G
BLDPBD7679-5G
BLDPBD7679-25G
2-Bromo-4'-(methylsulphonyl)acetophenone 98%
2-Bromo-4'-(methylsulphonyl)acetophenone
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₉H₉BrO₃S MW: 277,1350 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00673134 Núm. CAS: 50413-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...