2-Hydroxy-5-methylisophthalaldehyde 94%
|
|
BLDPBD7626-5GEA
170
EUR
BLDPBD7626-5G
BLDPBD7626-1G
BLDPBD7626-25G
BLDPBD7626-250MG
BLDPBD7626-10G
2-Hydroxy-5-methylisophthalaldehyde 94%
2-Hydroxy-5-methylisophthalaldehyde
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₉H₈O₃ MW: 164,1600 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00016596 Núm. CAS: 7310-95-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...