2-Hydroxy-3-methylbenzhydrazide 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD75567-1GEA 40 EUR
BLDPBD75567-1G BLDPBD75567-250MG BLDPBD75567-5G
2-Hydroxy-3-methylbenzhydrazide 98%
2-Hydroxy-3-methylbenzhydrazide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₁₀N₂O₂
MW: 166,18 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00017052
Núm. CAS: 30991-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR