3-(Trifluoromethoxy)salicylaldehyde 98+%

Proveedor: BLD PHARMATECH GMBH

BLDPBD75529-1GEA 88 EUR
BLDPBD75529-1G BLDPBD75529-250MG BLDPBD75529-100MG
3-(Trifluoromethoxy)salicylaldehyde 98+%
3-(Trifluoromethoxy)salicylaldehyde

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₈H₅F₃O₃
MW: 206,12 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD03788527
Núm. CAS: 497959-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR