Tetrahydro-2H-thiopyran-4-carboxylic acid 98%

BLDPBD75437-25GEA 586 EUR
BLDPBD75437-25G BLDPBD75437-1G BLDPBD75437-5G
Tetrahydro-2H-thiopyran-4-carboxylic acid 98%
Tetrahydro-2H-thiopyran-4-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₁₀O₂S
MW: 146,207 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09879921
Núm. CAS: 89489-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR