2-Fluoro-4-iodobenzoic acid 98%
|
|
BLDPBD74873-100GEA
621
EUR
BLDPBD74873-100G
BLDPBD74873-100MG
BLDPBD74873-10G
BLDPBD74873-250MG
BLDPBD74873-25G
BLDPBD74873-5G
BLDPBD74873-1G
2-Fluoro-4-iodobenzoic acid 98%
2-Fluoro-4-iodobenzoic acid
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₇H₄FIO₂ MW: 266,0100 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03094173 Núm. CAS: 124700-40-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...