Isoquinolin-N-oxide 97%

BLDPBD7460-1GEA 15 EUR
BLDPBD7460-1G
Isoquinolin-N-oxide 97%
Isoquinolin-N-oxide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₇NO
MW: 145,16 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00006903
Núm. CAS: 1532-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR