4-Amino-2-(trifluoromethyl)pyridine 98%

BLDPBD74001-250MGEA 7 EUR
BLDPBD74001-250MG BLDPBD74001-100G BLDPBD74001-5G BLDPBD74001-1G BLDPBD74001-10G BLDPBD74001-25G
4-Amino-2-(trifluoromethyl)pyridine 98%
4-Amino-2-(trifluoromethyl)pyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₅F₃N₂
MW: 162,1100 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09056877
Núm. CAS: 147149-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR