2-Chloro-5-(trifluoromethyl)-4-iodopyridine 97%
|
|
BLDPBD73099-100GEA
246
EUR
BLDPBD73099-100G
BLDPBD73099-25G
BLDPBD73099-1G
BLDPBD73099-10G
BLDPBD73099-5G
2-Chloro-5-(trifluoromethyl)-4-iodopyridine 97%
2-Chloro-5-(trifluoromethyl)-4-iodopyridine
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₆H₂ClF₃IN MW: 307,44 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD01862099 Núm. CAS: 505084-55-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...