2-Butylimidazole 97%

BLDPBD72413-100GEA 283 EUR
BLDPBD72413-100G BLDPBD72413-1G BLDPBD72413-5G BLDPBD72413-25G
2-Butylimidazole 97%
2-Butylimidazole

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₁₂N₂
MW: 124,1800 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00792664
Núm. CAS: 50790-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR