2,4-Dichlorocinnamic acid 97%

BLDPBD7236-10GEA 24 EUR
BLDPBD7236-10G BLDPBD7236-5G BLDPBD7236-25G BLDPBD7236-100G
2,4-Dichlorocinnamic acid 97%
2,4-Dichlorocinnamic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₆Cl₂O₂
MW: 217,0487 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00004373
Núm. CAS: 1201-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR