2-Bromo-N-methylbenzamide 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD72251-5GEA 197 EUR
BLDPBD72251-5G BLDPBD72251-250MG BLDPBD72251-1G
2-Bromo-N-methylbenzamide 98%
2-Bromo-N-methylbenzamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₈BrNO
MW: 214,06 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00099250
Núm. CAS: 61436-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR