4-Bromo-1-chloroisoquinoline 97%

BLDPBD7170-250MGEA 10 EUR
BLDPBD7170-0.025G BLDPBD7170-250MG BLDPBD7170-1G BLDPBD7170-5G BLDPBD7170-100MG BLDPBD7170-25G
4-Bromo-1-chloroisoquinoline 97%
4-Bromo-1-chloroisoquinoline

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₅BrClN
MW: 242,4997 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00234469
Núm. CAS: 66728-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR