2-Bromo-3-fluorobenzaldehyde 98%
|
|
BLDPBD71598-25GEA
112
EUR
BLDPBD71598-25G
BLDPBD71598-10G
BLDPBD71598-5G
BLDPBD71598-500G
BLDPBD71598-100G
BLDPBD71598-1G
2-Bromo-3-fluorobenzaldehyde 98%
2-Bromo-3-fluorobenzaldehyde
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₇H₄BrFO MW: 203,01 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD07782044 Núm. CAS: 891180-59-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...