2-Aminopyridine-N-oxide 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD70960-250MGEA 61 EUR
BLDPBD70960-250MG BLDPBD70960-1G BLDPBD70960-100MG
2-Aminopyridine-N-oxide 98%
2-Aminopyridine-N-oxide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₅H₆N₂O
MW: 110,1139 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00059168
Núm. CAS: 14150-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR