2-Amino-4,6-dibromopyrimidine 97%

BLDPBD70039-10GEA 675 EUR
BLDPBD70039-10G BLDPBD70039-250MG BLDPBD70039-1G BLDPBD70039-5G
2-Amino-4,6-dibromopyrimidine 97%
2-Amino-4,6-dibromopyrimidine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₄H₃Br₂N₃
MW: 252,8947 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD09263251
Núm. CAS: 856973-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR