2-Acetylamino-5-bromo-6-methylpyridine 98%

BLDPBD69480-1GEA 62 EUR
BLDPBD69480-1G BLDPBD69480-250MG
2-Acetylamino-5-bromo-6-methylpyridine 98%
2-Acetylamino-5-bromo-6-methylpyridine

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₉BrN₂O
MW: 229,0700 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD03909755
Núm. CAS: 142404-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR