4-[3-(Dimethylamino)propoxy]benzeneboronic acid, pinacol ester 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD69246-5GEA 356 EUR
BLDPBD69246-5G BLDPBD69246-100MG BLDPBD69246-1G BLDPBD69246-250MG
4-[3-(Dimethylamino)propoxy]benzeneboronic acid, pinacol ester 97%
4-[3-(Dimethylamino)propoxy]benzeneboronic acid, pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₇H₂₈BNO₃
MW: 305,22 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD09702395
Núm. CAS: 627899-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR