2,8-Dichloroquinoline 98%
|
|
BLDPBD68682-250MGEA
7
EUR
BLDPBD68682-250MG
BLDPBD68682-25G
BLDPBD68682-10G
BLDPBD68682-1G
BLDPBD68682-5G
2,8-Dichloroquinoline 98%
2,8-Dichloroquinoline
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₉H₅Cl₂N MW: 198,0500 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00832077 Núm. CAS: 4470-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...