Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')palladium(II) 98%

BLDPBD682428-1GEA 149 EUR
BLDPBD682428-1G BLDPBD682428-250MG BLDPBD682428-100MG
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')palladium(II) 98%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')palladium(II)

Storage condition: Inert atmosphere, 2-8°C

Formula: C₂₂H₃₈O₄Pd
MW: 472,9600 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD16038137
Núm. CAS: 15214-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR