2,6-Dichloroquinoline 97%

Proveedor: BLD PHARMATECH GMBH

BLDPBD68240-5GEA 69 EUR
BLDPBD68240-5G BLDPBD68240-250MG BLDPBD68240-1G BLDPBD68240-25G BLDPBD68240-100G BLDPBD68240-10G
2,6-Dichloroquinoline 97%
2,6-Dichloroquinoline

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₅Cl₂N
MW: 198,0500 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD03427194
Núm. CAS: 1810-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR