4,6-Dimethoxysalicylaldehyde 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD6729-1GEA 17 EUR
BLDPBD6729-1G BLDPBD6729-10G BLDPBD6729-250MG BLDPBD6729-100MG BLDPBD6729-100G BLDPBD6729-5G BLDPBD6729-25G
4,6-Dimethoxysalicylaldehyde 98%
4,6-Dimethoxysalicylaldehyde

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₉H₁₀O₄
MW: 182,1733 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00003328
Núm. CAS: 708-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR