5-Nitro-2,4-xylidine 97%
Proveedor: BLD PHARMATECH GMBH
|
|
BLDPBD67072-250MGEA
9
EUR
BLDPBD67072-250MG
BLDPBD67072-10G
BLDPBD67072-1G
BLDPBD67072-25G
BLDPBD67072-5G
BLDPBD67072-100G
5-Nitro-2,4-xylidine 97%
5-Nitro-2,4-xylidine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₈H₁₀N₂O₂ MW: 166,1800 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00025199 Núm. CAS: 2124-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...