2-(2-Hydroxyethoxy)benzaldehyde 96%

Proveedor: BLD PHARMATECH GMBH

BLDPBD6681-5GEA 18 EUR
BLDPBD6681-5G BLDPBD6681-25G BLDPBD6681-100G
2-(2-Hydroxyethoxy)benzaldehyde 96%
2-(2-Hydroxyethoxy)benzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₁₀O₃
MW: 166,1739 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00191449
Núm. CAS: 22042-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR