2,4,6-Tribromobenzoic acid 95%

Proveedor: BLD PHARMATECH GMBH

BLDPBD66193-1GEA 74 EUR
BLDPBD66193-1G BLDPBD66193-25G BLDPBD66193-5G BLDPBD66193-250MG
2,4,6-Tribromobenzoic acid 95%
2,4,6-Tribromobenzoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₃Br₃O₂
MW: 358,8095 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00059882
Núm. CAS: 633-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR