5-(Ethoxycarbonyl)-2-fluorophenylboronic acid 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD6577-1GEA 51 EUR
BLDPBD6577-1G BLDPBD6577-5G BLDPBD6577-0.025G BLDPBD6577-25G BLDPBD6577-250MG
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid 98%
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₁₀BFO₄
MW: 211,98 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD08235094
Núm. CAS: 874219-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR