2,3-Dihydro-1-benzofuran-5-sulphonoylchloride 96%

BLDPBD65629-25GEA 301 EUR
BLDPBD65629-25G BLDPBD65629-250MG BLDPBD65629-10G BLDPBD65629-5G BLDPBD65629-1G
2,3-Dihydro-1-benzofuran-5-sulphonoylchloride 96%
2,3-Dihydro-1-benzofuran-5-sulphonoylchloride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₇ClO₃S
MW: 218,66 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD02677703
Núm. CAS: 115010-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR