N-Phenylglycine 98%
|
|
BLDPBD6524-5GEA
13
EUR
BLDPBD6524-5G
BLDPBD6524-25G
BLDPBD6524-100G
N-Phenylglycine 98%
N-Phenylglycine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₈H₉NO₂ MW: 151,17 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00014009 Núm. CAS: 103-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...