5-Fluoro-(2-methoxycarbonyl)phenylboronic acid 98%

Proveedor: BLD PHARMATECH GMBH

BLDPBD6259-100MGEA 18 EUR
BLDPBD6259-100MG BLDPBD6259-5G BLDPBD6259-250MG BLDPBD6259-1G
5-Fluoro-(2-methoxycarbonyl)phenylboronic acid 98%
5-Fluoro-(2-methoxycarbonyl)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₈BFO₄
MW: 197,96 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD03095367
Núm. CAS: 850568-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR