4-Methoxy-3-(trifluoromethyl)phenylboronic acid 98%
|
|
BLDPBD6251-1GEA
48
EUR
BLDPBD6251-1G
BLDPBD6251-10G
BLDPBD6251-250MG
BLDPBD6251-5G
4-Methoxy-3-(trifluoromethyl)phenylboronic acid 98%
4-Methoxy-3-(trifluoromethyl)phenylboronic acid
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₈H₈BF₃O₃ MW: 219,95 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD07363790 Núm. CAS: 149507-36-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...