2-(4-Bromophenyl)imidazo[1,2-a]pyridine 97%

BLDPBD62108-250MGEA 18 EUR
BLDPBD62108-250MG BLDPBD62108-5G BLDPBD62108-1G
2-(4-Bromophenyl)imidazo[1,2-a]pyridine 97%
2-(4-Bromophenyl)imidazo[1,2-a]pyridine

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₃H₉BrN₂
MW: 273,13 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00218250
Núm. CAS: 34658-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR