4-Amino-3-chloro-5-methylbenzonitrile 98%
|
|
BLDPBD6081-5GEA
154
EUR
BLDPBD6081-5G
BLDPBD6081-1G
BLDPBD6081-250MG
4-Amino-3-chloro-5-methylbenzonitrile 98%
4-Amino-3-chloro-5-methylbenzonitrile
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₈H₇ClN₂ MW: 166,61 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00041476 Núm. CAS: 158296-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...