4'-Amino-3',5'-dichloroacetophenone 98%

BLDPBD6073-1GEA 7 EUR
BLDPBD6073-1G BLDPBD6073-25G BLDPBD6073-5G BLDPBD6073-10G BLDPBD6073-100G
4'-Amino-3',5'-dichloroacetophenone 98%
4'-Amino-3',5'-dichloroacetophenone

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₈H₇Cl₂NO
MW: 204,0500 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00238535
Núm. CAS: 37148-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR