1-Boc-1H-pyrazole-4-boronic acid pinacol ester 98%

BLDPBD60461-250MGEA 6 EUR
BLDPBD60461-250MG BLDPBD60461-100G BLDPBD60461-10G BLDPBD60461-500G BLDPBD60461-0.025G BLDPBD60461-1G BLDPBD60461-25G BLDPBD60461-5G
1-Boc-1H-pyrazole-4-boronic acid pinacol ester 98%
1-Boc-1H-pyrazole-4-boronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₄H₂₃N₂O₄B
MW: 294,1544 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD05663873
Núm. CAS: 552846-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR