1-Pyrenecarboxylic acid 97%
|
|
BLDPBD60364-25GEA
1024
EUR
BLDPBD60364-25G
BLDPBD60364-250MG
BLDPBD60364-5G
BLDPBD60364-100MG
BLDPBD60364-1G
1-Pyrenecarboxylic acid 97%
1-Pyrenecarboxylic acid
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₇H₁₀O₂ MW: 246,2601 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00192424 Núm. CAS: 19694-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...