1-Monolinoleoyl-rac-glycerol 98%

BLDPBD59687-100MGEA 150 EUR
BLDPBD59687-100MG
1-Monolinoleoyl-rac-glycerol 98%
1-Monolinoleoyl-rac-glycerol

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formula: C₂₁H₃₈O₄
MW: 354,531 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00270409
Núm. CAS: 2277-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR