4-Methoxythioanisole 97%

BLDPBD59258-25GEA 97 EUR
BLDPBD59258-25G BLDPBD59258-100G BLDPBD59258-5G BLDPBD59258-1G
4-Methoxythioanisole 97%
4-Methoxythioanisole

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₁₀OS
MW: 154,2294 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00010587
Núm. CAS: 1879-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR