1H-Pyrazole-4-boronic acid 95%

Proveedor: BLD PHARMATECH GMBH

BLDPBD58914-1GEA 11 EUR
BLDPBD58914-1G BLDPBD58914-250MG BLDPBD58914-25G BLDPBD58914-5G BLDPBD58914-10G
1H-Pyrazole-4-boronic acid 95%
1H-Pyrazole-4-boronic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₃H₅BN₂O₂
MW: 111,9000 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD07368251
Núm. CAS: 763120-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR