4-(Propan-2-yl)-1H-pyrazol-3-amine 95%

BLDPBD58891-100MGEA 23 EUR
BLDPBD58891-100MG BLDPBD58891-1G BLDPBD58891-5G BLDPBD58891-250MG
4-(Propan-2-yl)-1H-pyrazol-3-amine 95%
4-(Propan-2-yl)-1H-pyrazol-3-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₁₁N₃
MW: 125,172 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD11112127
Núm. CAS: 151521-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR